missing translation for 'onlineSavingsMsg'
Learn More

Di-p-toluoyl-D-tartaric Acid Monohydrate, TRC

Product Code. 30136391
Change view
Click to view available options
Quantity:
2.5 g
5 g
Unit Size:
2.5g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136391 5 g 5g
17974315 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136391 Supplier Toronto Research Chemicals Supplier No. TRCD4943885G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71607-31-3
Molecular Formula C20H20O9
Molecular Weight (g/mol) 404.37
Synonym (2S,3S)-2,3-Bis((4-Methylbenzoyl)oxy)succinic Acid Hydrate
IUPAC Name (2S,3S)-2,3-bis((4-methylbenzoyl)oxy)succinic acid hydrate
SMILES CC1=CC=C(C(O[C@@H]([C@H](OC(C2=CC=C(C)C=C2)=O)C(O)=O)C(O)=O)=O)C=C1.O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C20H18O8.H2O/c1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14;/h3-10,15-16H,1-2H3,(H,21,22)(H,23,24);1H2/t15-,16-;/m0./s1
Formula Weight 404.11
Chemical Name or Material Di-p-toluoyl-D-tartaric Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.