missing translation for 'onlineSavingsMsg'
Learn More

(+)-Di-p-toluoyl-D-(+)-tartaric Acid, Mikromol™

Product Code. 17971906
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971906 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971906 Supplier Mikromol Supplier No. MM1497.030025
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(+)-Di-p-toluoyl-D-(+)-tartaric Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (+)-O,O'-Di-p-toluoyl-D-tartaric Acid
CAS 32634-68-7
Analyte or Component Names (+)-O,O'-Di-p-toluoyl-D-tartaric Acid
Shipping Condition Room Temperature
Molecular Formula C20 H18 O8
Certifications/Compliance ISO 17025
Synonym (2S,3S)-2,3-Bis[(4-methylbenzoyl)oxy]butanedioic acid, (+)-Di-p-toluoyl-D-(+)-tartaric Acid, Butanedioic acid, 2,3-bis[(4-methylbenzoyl)oxy]-, (2S,3S)-, Butanedioic acid, 2,3-bis[(4-methylbenzoyl)oxy]-, [S-(R*,R*)]-, Tartaric acid, di-p-toluate, (+)- (8CI), p-Toluic acid, diester with D-tartaric acid (7CI), (+)-Bis-O,O'-(4-toluoyl)-D-tartrate, (+)-Di-O,O'-p-toluoyltartaric acid, (+)-Di-O,O'-p-toluyltartaric acid, (+)-Di-O-(p-toluoyl)tartaric acid, (+)-Di-O-p-toluoyl-D-tartaric acid, (+)-Di-p-toluoyl-D-tartaric acid, (+)-Di-p-toluoyltartaric acid, (+)-Ditoluoyltartaric acid, (+)-O,O-Di-p-toluoyltartaric acid, (2S,3S)-(+)-Di-p-toluoyltartaric acid, (2S,3S)-2,3-Bis[(4-methylbenzoyl)oxy]succinic acid, (S,S)-O,O'-Di-p-toluoyltartaric acid, D-Bis(O-4-methylbenzoyl)tartaric acid, D-Di-O,O'-p-toluyltartaric acid, D-Di-p-toluoyltartaric acid, Di-p-toluoyl-D-tartaric acid, Di-p-toluoyl-l-tartaric acid, NSC 97592, Rivastigmine Related Compound A
InChI Formula InChI=1S/C20H18O8/c1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14/h3-10,15-16H,1-2H3,(H,21,22)(H,23,24)/t15-,16-/m0/s1
SMILES Cc1ccc(cc1)C(=O)O[C@@H]([C@H](OC(=O)c2ccc(C)cc2)C(=O)O)C(=O)O
IUPAC Name (2S,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid
Molecular Weight (g/mol) 386.35
Quantity 25 mg
Formula Weight 386.1002 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.