missing translation for 'onlineSavingsMsg'
Learn More

Di-p-toluoyl-D-tartaric acid, 98%

Product Code. p-7058394
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11317976

Brand: Thermo Scientific Alfa Aesar L13223.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 32634-68-7 | C20H18O8 | 386.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32634-68-7
Molecular Formula C20H18O8
Molecular Weight (g/mol) 386.36
MDL Number MFCD00008552
InChI Key CMIBUZBMZCBCAT-HOTGVXAUSA-N
Synonym di-p-toluoyl-d-tartaric acid, +-di-1,4-toluoyl-d-tartaric acid, 2s,3s-2,3-bis 4-methylbenzoyl oxy succinic acid, unii-4480ma5qip, ditoluoyltartaric acid, +, +-dtta, butanedioic acid, 2,3-bis 4-methylbenzoyl oxy-, 2s,3s, 2s,3s-2,3-bis 4-methylbenzoyl oxy butanedioic acid, --di-p-toluoyl-l-trataric acid
PubChem CID 263211
SMILES CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 169°C to 171°C
Beilstein 3225584
IUPAC Name (2S,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid
Formula Weight 386.36
Percent Purity 98%
Quantity 5 g
Optical Rotation +139° (c=1 in Ethanol)
Chemical Name or Material Di-p-toluoyl-D-tartaric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.