missing translation for 'onlineSavingsMsg'
Learn More

Di-p-tert-Butylphenyl Phosphorochloridate, TRC

Product Code. 30380431
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380431 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380431 Supplier Toronto Research Chemicals Supplier No. TRCD4285352.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-p-tert-Butylphenyl Phosphorochloridate
CAS 38135-31-8
Molecular Formula C20H26ClO3P
Quantity 2.5 g
Synonym Bis[4-(1,1-dimethylethyl)phenyl] Ester Phosphorochloridic Acid
InChI Formula InChI=1S/C20H26ClO3P/c1-19(2,3)15-7-11-17(12-8-15)23-25(21,22)24-18-13-9-16(10-14-18)20(4,5)6/h7-14H,1-6H3
SMILES O=P(OC1=CC=C(C(C)(C)C)C=C1)(Cl)OC2=CC=C(C(C)(C)C)C=C2
IUPAC Name bis(4-(tert-butyl)phenyl) phosphorochloridate
Molecular Weight (g/mol) 380.85
Formula Weight 380.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.