missing translation for 'onlineSavingsMsg'
Learn More

Di-o-tolyl-phosphate, TRC

Product Code. 17963655
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963655 1 g 1g
17940605 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17963655 Supplier Toronto Research Chemicals Supplier No. TRCD4945751G
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35787-74-7
Molecular Formula C14H15O4P
Molecular Weight (g/mol) 278.24
Synonym Di-o-Tolyl Phosphate, o-Tolyl phosphate, (C7H7O)2(HO)PO (7CI), Di-o-cresyl hydrogen phosphate, Di-o-cresyl phosphate, Di-o-tolyl phosphate, Phosphoric acid di-o-tolyl ester, Bis(2-methylphenyl) Ester Phosphoric Acid, Di-o-Cresyl Hydrogen Phosphate, Di-o-cresyl Phosphate, Di-o-tolyl Phosphate, Phosphoric Acid Di-o-tolyl Ester
IUPAC Name bis(2-methylphenyl) hydrogen phosphate
SMILES Cc1ccccc1OP(=O)(O)Oc2ccccc2C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H15O4P/c1-11-7-3-5-9-13(11)17-19(15,16)18-14-10-6-4-8-12(14)2/h3-10H,1-2H3,(H,15,16)
Formula Weight 278.0708
Percent Purity >95%
Chemical Name or Material Di-o-tolyl-phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.