missing translation for 'onlineSavingsMsg'
Learn More

Di-O-glycinoylcurcumin Dihydrochloride, TRC

Product Code. 30385227
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385227 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385227 Supplier Toronto Research Chemicals Supplier No. TRCD455700100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-O-glycinoylcurcumin Dihydrochloride
CAS 1322544-82-0
Molecular Formula C25H28Cl2N2O8
Quantity 100 mg
Synonym ((1E,6E)-3,5-Dioxohepta-1,6-diene-1,7-diyl)bis(2-methoxy-4,1-phenylene) bis(2-aminoacetate) Dihydrochloride
InChI Formula InChI=1S/C25H26N2O8.2ClH/c1-32-22-11-16(5-9-20(22)34-24(30)14-26)3-7-18(28)13-19(29)8-4-17-6-10-21(23(12-17)33-2)35-25(31)15-27;;/h3-12H,13-15,26-27H2,1-2H3;2*1H/b7-3+,8-4+;;
SMILES O=C(CC(/C=C/C1=CC(OC)=C(OC(CN)=O)C=C1)=O)/C=C/C2=CC=C(OC(CN)=O)C(OC)=C2
IUPAC Name ((1E,6E)-3,5-dioxohepta-1,6-diene-1,7-diyl)bis(2-methoxy-4,1-phenylene) bis(2-aminoacetate) dihydrogen chloride
Molecular Weight (g/mol) 555.4
Formula Weight 554.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.