missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propylamine, 99%

Product Code. 11342787
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342787 500 mL 500mL
11332787 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11342787 Supplier Thermo Scientific Alfa Aesar Supplier No. L15808.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 142-84-7 | C6H15N | 101.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-84-7
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00009362
InChI Key WEHWNAOGRSTTBQ-UHFFFAOYSA-N
Synonym dipropylamine, di-n-propylamine, 1-propanamine, n-propyl, n-dipropylamine, n-propyl-1-propanamine, rcra waste number u110, dipropyl amine, rcra waste no. u110, unii-60p318iiry, ccris 4805
PubChem CID 8902
IUPAC Name N-propylpropan-1-amine
SMILES CCCNCCC

Specifications

Melting Point -63°C
Density 0.738
Boiling Point 109°C to 110°C
Flash Point 7°C (45°F)
Odor Ammonia-like
Refractive Index 1.405
Quantity 500 mL
UN Number UN2383
Beilstein 505974
Merck Index 14,3343
Formula Weight 101.19
Percent Purity 99%
Chemical Name or Material Di-n-propylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.