missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl sulfite, 99%, Thermo Scientific™

Product Code. 11368575
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368575 25 g 25g
11358575 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368575 Supplier Thermo Scientific Alfa Aesar Supplier No. L12033.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-98-3
Molecular Formula C6H14O3S
Molecular Weight (g/mol) 166.235
MDL Number MFCD00059404
InChI Key MAIQPVFXODAAIG-UHFFFAOYSA-N
Synonym di-n-propyl sulfite, di-n-propyl sulphite, sulfurous acid, dipropyl ester, dipropyl sulphite, dipropylsulfite, di-n-propylsulfit, sulfurous acid dipropyl ester, sulphurous acid dipropyl ester, dipropyl ester of sulfurous acid
PubChem CID 136434
IUPAC Name dipropyl sulfite
SMILES CCCOS(=O)OCCC

Specifications

Density 1.03
Boiling Point 107°C to 109°C (42 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.421
Quantity 25 g
UN Number UN3334
Beilstein 1703571
Sensitivity Moisture sensitive
Formula Weight 166.24
Percent Purity 99%
Chemical Name or Material Di-n-propyl sulfite
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.