missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl phthalate, 98%

Product Code. 11446976
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11446976

Brand: Thermo Scientific Alfa Aesar B24056.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 131-16-8 | C14H18O4 | 250.294 g/mol

Di-n-propyl phthalate is used to make plasticizers and polymer additives. It is also used in chemical reagents, organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-propyl phthalate is used to make plasticizers and polymer additives. It is also used in chemical reagents, organic intermediates.

Solubility
Soluble in chloroform, methanol.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-16-8
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.294
MDL Number MFCD00009371
InChI Key MQHNKCZKNAJROC-UHFFFAOYSA-N
Synonym dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss
PubChem CID 8559
ChEBI CHEBI:60069
IUPAC Name dipropyl benzene-1,2-dicarboxylate
SMILES CCCOC(=O)C1=CC=CC=C1C(=O)OCCC

Specifications

Density 1.082
Boiling Point 168°C to 170°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.497
Quantity 250 g
UN Number UN3082
Beilstein 2332522
Solubility Information Soluble in chloroform,methanol.
Formula Weight 250.3
Percent Purity 98%
Chemical Name or Material Di-n-propyl phthalate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.