Learn More
Di-n-propyl ether, 99%
Description
Di-n-propyl ether, is used as a solvent and intermediate for organic chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Di-n-propyl ether, is used as a solvent and intermediate for organic chemical synthesis.
Solubility
It is slightly soluble in water 3 g/L (20°C), soluble in alcohol and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids, halogens.
Specifications
Specifications
| Melting Point | -122°C |
| Density | 0.747 |
| Boiling Point | 90°C to 91°C |
| Flash Point | −28°C (−18°F) |
| Quantity | 25 g |
| UN Number | UN2384 |
| Solubility Information | It is slightly soluble in water 3g/L (20°C),soluble in alcohol and ether. |
| Formula Weight | 102.18 |
| Percent Purity | 99% |
| Chemical Name or Material | Di-n-propyl ether |
Safety and Handling
- Di-n-propyl ether
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- MIXTURE LIST-Contain: Dipropyl ether
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.