missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyldichlorosilane, 97%

Product Code. 11379218
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379218 10 g 10g
11369218 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379218 Supplier Thermo Scientific Alfa Aesar Supplier No. 042599.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18416-07-4 | C16H34Cl2Si | 325.433 g/mol

Di-n-octyldichlorosilane is used in the synthesis of organic photovoltaics. It is also used in medicine field, electronic industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18416-07-4
Molecular Formula C16H34Cl2Si
Molecular Weight (g/mol) 325.433
MDL Number MFCD01074472
InChI Key CVAGYCLEIYGJQT-UHFFFAOYSA-N
Synonym di-n-octyldichlorosilane, dichloro dioctyl silane, silane, dichlorodioctyl, dioctyldichlorosilane, dioctyldichlorsilan, dichlorodi-n-octylsilane, di-n-octyldichlorosilane 10g
PubChem CID 248429
IUPAC Name dichloro(dioctyl)silane
SMILES CCCCCCCC[Si](CCCCCCCC)(Cl)Cl

Specifications

Boiling Point 145°C (0.2mmHg)
Color Colorless to Yellow
Physical Form Liquid
Quantity 10 g
Linear Formula [CH3(CH2)7]2SiCl2
UN Number UN2987
Solubility Information Sparingly Soluble (8.8E-6g/L) at 25°C.
Formula Weight 325.44
Percent Purity 97%
Sensitivity Moisture sensitive
Density 0.940 g/mL
Chemical Name or Material Di-n-octyldichlorosilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.