missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyldichlorosilane, 97%, Thermo Scientific Chemicals

Catalog Number p-7052124
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
18416-07-4
C16H34Cl2Si
325.433
MFCD01074472
CVAGYCLEIYGJQT-UHFFFAOYSA-N
di-n-octyldichlorosilane, dichloro dioctyl silane, silane, dichlorodioctyl, dioctyldichlorosilane, dioctyldichlorsilan, dichlorodi-n-octylsilane, di-n-octyldichlorosilane 10g
248429
dichloro(dioctyl)silane
CCCCCCCC[Si](CCCCCCCC)(Cl)Cl
This item is not returnable. View return policy
18416-07-4
C16H34Cl2Si
325.433
MFCD01074472
CVAGYCLEIYGJQT-UHFFFAOYSA-N
di-n-octyldichlorosilane, dichloro dioctyl silane, silane, dichlorodioctyl, dioctyldichlorosilane, dioctyldichlorsilan, dichlorodi-n-octylsilane, di-n-octyldichlorosilane 10g
248429
dichloro(dioctyl)silane
CCCCCCCC[Si](CCCCCCCC)(Cl)Cl

Di-n-octyldichlorosilane is used in the synthesis of organic photovoltaics. It is also used in medicine field, electronic industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 145°C (0.2mmHg)
Color Colorless to Yellow
Physical Form Liquid
Quantity 10 g
Linear Formula [CH3(CH2)7]2SiCl2
UN Number UN2987
Solubility Information Sparingly Soluble (8.8E-6g/L) at 25°C.
Formula Weight 325.44
Percent Purity 97%
Sensitivity Moisture sensitive
Density 0.940 g/mL
Chemical Name or Material Di-n-octyldichlorosilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'