missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octylamine, 96%

Product Code. 11429156
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429156 25 g 25g
11439156 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429156 Supplier Thermo Scientific Alfa Aesar Supplier No. A18017.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1120-48-5 | C16H35N | 241.463 g/mol

Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.

Solubility
Immiscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-48-5
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.463
MDL Number MFCD00009557
InChI Key LAWOZCWGWDVVSG-UHFFFAOYSA-N
Synonym dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine
PubChem CID 3094
IUPAC Name N-octyloctan-1-amine
SMILES CCCCCCCCNCCCCCCCC

Specifications

Melting Point 13°C to 15°C
Density 0.799
Boiling Point 297°C to 298°C
Flash Point 142°C (287°F)
Odor Amine-like
Linear Formula [CH3(CH2)7]2NH
Refractive Index 1.4432
Quantity 25 g
UN Number UN2735
Beilstein 1748376
Solubility Information Immiscible in water.
Formula Weight 241.46
Percent Purity 98%
Chemical Name or Material Di-n-octylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-n-octylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Di-n-octylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.