missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-octyl Phthalate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-n-octyl ester-3,4,5,6-D4 |
| CAS | 93952-13-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 2H4 H34 O4 |
| Quantity | 10 mg |
| Synonym | Di-n-octyl Phthalate-d4, Phthalic acid bis-n-octyl ester-3,4,5,6 D4, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dioctyl ester (9CI), Phthalic Acid Di-n-octyl Ester (3,4,5,6)-D4, Phthalic Acid, Dioctyl Ester D4 (phenyl D4) |
| InChI Formula | InChI=1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3/i13D,14D,17D,18D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OCCCCCCCC)c(C(=O)OCCCCCCCC)c1[2H] |
| IUPAC Name | dioctyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 394.58 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?