missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl phenylphosphonate, 97%

Product Code. 11479040
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479040 100 g 100g
11459040 5 g 5g
11469040 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479040 Supplier Thermo Scientific Alfa Aesar Supplier No. A13468.22
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1754-47-8 | C22H39O3P | 382.53 g/mol

Di-n-octyl phenylphosphonate acts as a plasticizer, which is useful in ion-selective electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-octyl phenylphosphonate acts as a plasticizer, which is useful in ion-selective electrodes.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1754-47-8
Molecular Formula C22H39O3P
Molecular Weight (g/mol) 382.53
MDL Number MFCD00009561
InChI Key HAKMAMKAFTZXOZ-UHFFFAOYSA-N
Synonym dioctyl phenylphosphonate, di-n-octyl phenylphosphonate, dioctylphenylphosphonate, phenylphosphonic acid dioctyl ester, benzenephosphonic acid, dioctyl ester, di-n-octyl phenyl phosphonate, dioctyl phenyl phosphonate, phosphonic acid, phenyl-, dioctyl ester, phosphonic acid, p-phenyl-, dioctyl ester, di-n-octylphenylphosphonat
PubChem CID 15641
IUPAC Name dioctoxyphosphorylbenzene
SMILES CCCCCCCCOP(=O)(OCCCCCCCC)C1=CC=CC=C1

Specifications

Density 0.932
Boiling Point 219°C to 220°C (0.7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4795
Quantity 100 g
Beilstein 2948496
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 382.53
Percent Purity 97%
Chemical Name or Material Di-n-octyl phenylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.