missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octadecyl disulfide, 98%

Product Code. 11445097
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445097 5 g 5g
11435097 1 g 1g
11459447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445097 Supplier Thermo Scientific Alfa Aesar Supplier No. B21978.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2500-88-1 | C36H74S2 | 571.11 g/mol

It is used as a co stabilizer, antioxidant for plastics, polyoletins, HDPE, LDPE, PMMA, PA, engineering thermoplastic elastomers, synergist, thermal stabilizer, processing stabilizer in high temperature applications, peroxide decomposer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a co stabilizer, antioxidant for plastics, polyoletins, HDPE, LDPE, PMMA, PA, engineering thermoplastic elastomers, synergist, thermal stabilizer, processing stabilizer in high temperature applications, peroxide decomposer.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2500-88-1
Molecular Formula C36H74S2
Molecular Weight (g/mol) 571.11
MDL Number MFCD00026687
InChI Key MQQKTNDBASEZSD-UHFFFAOYSA-N
Synonym disulfide, dioctadecyl, dioctadecyl disulfide, octadecyl disulfide, di-n-octadecyl disulfide, distearyl disulfide, 1-octadecyldisulfanyl octadecane, dioctadecyl disulphide, di-n-octadecyldisulfide, dioctadecyldisulfid, n-octadecyl disulfide
PubChem CID 75627
IUPAC Name 1-(octadecyldisulfanyl)octadecane
SMILES CCCCCCCCCCCCCCCCCCSSCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 60°C to 64°C
Odor Unpleasant
Quantity 5 g
Beilstein 1715376
Solubility Information Insoluble in water.
Formula Weight 571.11
Percent Purity 98%
Chemical Name or Material Di-n-octadecyl disulfide

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.