missing translation for 'onlineSavingsMsg'
Learn More

Di-n-hexyl sulfide, 97%

Product Code. 11486026
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486026 100 g 100g
11476026 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486026 Supplier Thermo Scientific Alfa Aesar Supplier No. B22325.22
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6294-31-1 | C12H26S | 202.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6294-31-1
Molecular Formula C12H26S
Molecular Weight (g/mol) 202.40
MDL Number MFCD00009528
InChI Key LHNRHYOMDUJLLM-UHFFFAOYSA-N
Synonym hexyl sulfide, hexane, 1,1'-thiobis, dihexyl sulfide, di-n-hexyl sulfide, 7-thiatridecane, dihexyl sulphide, n-hexyl sulfide, di-n-hexylsulfide, hexylthiohexane, normal-hexyl sulfide
PubChem CID 80517
IUPAC Name 1-hexylsulfanylhexane
SMILES CCCCCCSCCCCCC

Specifications

Density 0.849
Boiling Point 260°C
Flash Point 116°C (241°F)
Odor Stench
Refractive Index 1.4587
Quantity 100 g
UN Number UN3334
Beilstein 1738333
Formula Weight 202.4
Percent Purity 97%
Chemical Name or Material Di-n-hexyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.