missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-hexyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-hexyl ester D4 |
| CAS | 1015854-55-3 |
| Molecular Formula | C20 D4 H26 O4 |
| Quantity | 0.1 g |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-dihexyl ester, Phthalic Acid Di-n-hexyl Ester (3,4,5,6) D4, Phthalic acid, bis-hexyl ester D4, Phthalic Acid, Dihexyl Ester D4 (phenyl D4) |
| InChI Formula | InChI=1 S/C20H30O4/c1-3-5-7-11-15-23-19(21)17-13-9-10-14-18(17)20(22)24-16-12-8-6-4-2/h9-10,13-14 H,3-8,11-12,15-16H2,1-2H3/i9D,10 D,13 D,14 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCC)c(C(=O)OCCCCCC)c1[2 H] |
| IUPAC Name | dihexyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 338.4744 |
| Formula Weight | 338.2395 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction