missing translation for 'onlineSavingsMsg'
Learn More

Di-n-hexyl ether, 98%

Product Code. 15418869
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418869 50 g 50g
15408869 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418869 Supplier Thermo Scientific Alfa Aesar Supplier No. L07076.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-58-3 | C12H26O | 186.34 g/mol

Dihexyl ether has been used as: supported liquid membrane during hollow-fiber liquid-phase microextraction method for determination of nitrophenolic compounds from atmospheric aerosol particles, an extraction solvent to detect avermectins in stream water by hollow-fiber-assisted liquid-phase microextraction technique coupled with LC-MS/MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dihexyl ether has been used as: supported liquid membrane during hollow-fiber liquid-phase microextraction method for determination of nitrophenolic compounds from atmospheric aerosol particles, an extraction solvent to detect avermectins in stream water by hollow-fiber-assisted liquid-phase microextraction technique coupled with LC-MS/MS.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-58-3
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00009525
InChI Key BPIUIOXAFBGMNB-UHFFFAOYSA-N
Synonym dihexyl ether, hexyl ether, hexane, 1,1'-oxybis, n-hexyl ether, di-n-hexyl ether, ether, dihexyl, bis 1-hexyl ether, 1,1'-oxybis hexane, 1-hexyloxy hexane
PubChem CID 8198
SMILES CCCCCCOCCCCCC

Specifications

Melting Point -43°C
Density 0.793
Boiling Point 228°C to 230°C
Flash Point 78°C (172°F)
Odor Mild
Linear Formula CH3(CH2)5O(CH2)5CH3
Refractive Index 1.4204
Quantity 50 g
Beilstein 1738177
Solubility Information Fully miscible in water.
IUPAC Name 1-(hexyloxy)hexane
Formula Weight 186.34
Percent Purity 98%
Chemical Name or Material Di-n-hexyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.