missing translation for 'onlineSavingsMsg'
Learn More

Di-n-heptyl Phthalate, TRC

Product Code. 30004448
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004448 1 g 1g
30006190 50 g 50g
30012536 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004448 Supplier Toronto Research Chemicals Supplier No. TRCD4464951G
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3648-21-3
Molecular Formula C22 H34 O4
Molecular Weight (g/mol) 362.5
Synonym Diheptyl phthalate, 1,2-Benzenedicarboxylic acid, 1,2-diheptyl ester (ACI), 1,2-Benzenedicarboxylic acid, diheptyl ester (9CI), Phthalic acid, diheptyl ester (6CI, 7CI, 8CI), Bis-heptyl phthalate, Di-n-heptyl phthalate, Heptyl phthalate, Sansocizer DHP
IUPAC Name diheptyl benzene-1,2-dicarboxylate
SMILES CCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H34O4/c1-3-5-7-9-13-17-25-21(23)19-15-11-12-16-20(19)22(24)26-18-14-10-8-6-4-2/h11-12,15-16H,3-10,13-14,17-18H2,1-2H3
Formula Weight 362.2457
Chemical Name or Material Phthalic acid,bis-n-heptyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.