missing translation for 'onlineSavingsMsg'
Learn More

Di-n-heptyl phthalate, 98+%

Product Code. 11492827
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492827 10 g 10g
11402837 50 g 50g
11412837 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492827 Supplier Thermo Scientific Alfa Aesar Supplier No. A15466.09
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3648-21-3 | C22H34O4 | 362.51 g/mol

Di-n-heptyl Phthalate is a Phthalate derivative and an organic extract contaminant found in drinking water, which was shown to activate Nrf-2-Mediated Antioxidant response in human cell line.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-heptyl Phthalate is a Phthalate derivative and an organic extract contaminant found in drinking water, which was shown to activate Nrf-2-Mediated Antioxidant response in human cell line.

Solubility
Sparingly soluble in water.

Notes
Incompatible with Strong acids, strong bases,.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3648-21-3
Molecular Formula C22H34O4
Molecular Weight (g/mol) 362.51
MDL Number MFCD00043680
InChI Key JQCXWCOOWVGKMT-UHFFFAOYSA-N
Synonym diheptyl phthalate, di-n-heptyl phthalate, heptyl phthalate, di-n-heptylphthalate, phthalic acid, diheptyl ester, phthalic acid diheptyl ester, 1,2-benzenedicarboxylic acid, diheptyl ester, n-diheptyl phthalate, unii-ie05vo8p8p, diheptyl ester of phthalic acid
PubChem CID 19284
ChEBI CHEBI:34677
IUPAC Name diheptyl benzene-1,2-dicarboxylate
SMILES CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC

Specifications

Density 0.992
Boiling Point 360°C
Flash Point 213°C (415°F)
Refractive Index 1.486
Quantity 10 g
Beilstein 2336623
Solubility Information Sparingly soluble in water.
Formula Weight 362.51
Percent Purity ≥98%
Chemical Name or Material Di-n-heptyl phthalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.