missing translation for 'onlineSavingsMsg'
Learn More

Di-n-decyldimethylammonium Bromide, TRC

Product Code. 30385019
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385019 100 mg
30375755 50 mg
30378737 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385019 Supplier Toronto Research Chemicals Supplier No. TRCD494853100MG
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-68-3
Molecular Formula C22H48BrN
Molecular Weight (g/mol) 406.53
SMILES [Br-].CCCCCCCCCC[N+](C)(C)CCCCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H48N.BrH/c1-5-7-9-11-13-15-17-19-21-23(3,4)22-20-18-16-14-12-10-8-6-2;/h5-22H2,1-4H3;1H/q+1;/p-1
Formula Weight 405.297
Chemical Name or Material Di-n-decyldimethylammonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.