missing translation for 'onlineSavingsMsg'
Learn More

Di-n-decyl sulfide, 97%

Product Code. 11435137
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435137 10 g 10g
11445137 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435137 Supplier Thermo Scientific Alfa Aesar Supplier No. B22560.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 693-83-4 | C20H42S | 314.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-83-4
Molecular Formula C20H42S
Molecular Weight (g/mol) 314.62
MDL Number MFCD00026542
InChI Key RKYMVQJWYYOIJB-UHFFFAOYSA-N
Synonym decyl sulfide, didecyl sulfide, di-n-decyl sulfide, decane, 1,1'-thiobis, n-decyl sulfide, 11-thiaheneicosane, decylthiodecane, didecyl sulphide, di-n-decylsulfide, decane,1'-thiobis
PubChem CID 69662
IUPAC Name 1-decylsulfanyldecane
SMILES CCCCCCCCCCSCCCCCCCCCC

Specifications

Melting Point 25°C to 28°C
Boiling Point 217°C to 218°C (8 mmHg)
Quantity 10 g
Beilstein 1763427
Formula Weight 314.63
Percent Purity 97%
Chemical Name or Material Di-n-decyl sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.