missing translation for 'onlineSavingsMsg'
Learn More

Di-n-decyl Phthalate-3,4,5,6-d4, TRC

Product Code. 30383655
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383655 2 mg 2mg
30374901 1 mg -
30377670 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30383655 Supplier Toronto Research Chemicals Supplier No. TRCD4394012MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1276197-18-2
Molecular Formula C28 2H4 H42 O4
Molecular Weight (g/mol) 450.6871
Synonym Phthalic Acid Didecyl Ester D4, Phthalic Acid, Didecyl Ester D4 (phenyl D4), Phthalic Acid, Bis-decyl Ester D4 (phenyl D4), Phthalic Acid Didecyl Ester D4, Phthalic Acid, Didecyl Ester D4 (phenyl D4), Phthalic Acid, Bis-decyl Ester D4 (phenyl D4)
IUPAC Name didecyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
SMILES [2H]c1c([2H])c([2H])c(C(=O)OCCCCCCCCCC)c(C(=O)OCCCCCCCCCC)c1[2H]

Specifications

Quantity 2 mg
InChI Formula InChI=1S/C28H46O4/c1-3-5-7-9-11-13-15-19-23-31-27(29)25-21-17-18-22-26(25)28(30)32-24-20-16-14-12-10-8-6-4-2/h17-18,21-22H,3-16,19-20,23-24H2,1-2H3/i17D,18D,21D,22D
Formula Weight 450.3647
Chemical Name or Material Phthalic Acid Didecyl Ester D4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.