missing translation for 'onlineSavingsMsg'
Learn More

Di-n-decyl dimethylammonium bromide, 80% aq. gel

Product Code. 11309879
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309879 100 g 100g
11319879 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309879 Supplier Thermo Scientific Alfa Aesar Supplier No. B24832.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2390-68-3 | C22H48BrN | 406.537 g/mol

Di-n-decyl dimethylammonium bromide is an efficient phase-transfer catalyst useful in many organic reactions. It is useful in many organic transformations such as isomerization, esterification, and alkylation among others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-decyl dimethylammonium bromide is an efficient phase-transfer catalyst useful in many organic reactions. It is useful in many organic transformations such as isomerization, esterification, and alkylation among others.

Solubility
Soluble in methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-68-3
Molecular Formula C22H48BrN
Molecular Weight (g/mol) 406.537
MDL Number MFCD00012194
InChI Key UMGXUWVIJIQANV-UHFFFAOYSA-M
Synonym didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide
PubChem CID 16957
IUPAC Name didecyl(dimethyl)azanium;bromide
SMILES CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-]

Specifications

Linear Formula [CH3(CH2)9]2N(CH3)2Br
Refractive Index 1.457
Quantity 100 g
Solubility Information Soluble in methanol.
Formula Weight 406.54
Chemical Name or Material Di-n-decyl dimethylammonium bromide, 80% aq. gel

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.