missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin oxide, 98%

Product Code. 11363546
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11363546

Brand: Thermo Scientific Alfa Aesar L14491.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 818-08-6 | C8H18OSn | 248.941 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 818-08-6
Molecular Formula C8H18OSn
Molecular Weight (g/mol) 248.941
MDL Number MFCD00001992
InChI Key JGFBRKRYDCGYKD-UHFFFAOYSA-N
Synonym dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
PubChem CID 61221
IUPAC Name dibutyl(oxo)tin
SMILES CCCC[Sn](=O)CCCC

Specifications

Melting Point >300°C
Odor Odorless
Linear Formula [CH3(CH2)3]2SnO
Quantity 25 g
UN Number UN3146
Beilstein 4126243
Formula Weight 248.94
Percent Purity 98%
Chemical Name or Material Di-n-butyltin oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.