missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin oxide

Product Code. 11397717
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397717 250 g 250g
11387717 50 g 50g
11377717 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397717 Supplier Thermo Scientific Alfa Aesar Supplier No. 014117.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 818-08-6 | C8H18OSn | 248.941 g/mol

Di-n-butyltin oxide is used as a reagent and a catalyst. It is particularly useful in regioselective alkylation, acylation and sulfonation reactions for starting materials containing alcohol functional groups. It is used in studies pertaining to discovering novel antitumor agents, antifungal activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 818-08-6
Molecular Formula C8H18OSn
Molecular Weight (g/mol) 248.941
MDL Number MFCD00001992
InChI Key JGFBRKRYDCGYKD-UHFFFAOYSA-N
Synonym dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
PubChem CID 61221
IUPAC Name dibutyl(oxo)tin
SMILES CCCC[Sn](=O)CCCC

Specifications

Melting Point >300°C
Color White
Density 1.58g/mL
Odor Odorless
Linear Formula [CH3(CH2)3]2SnO
Quantity 250 g
UN Number UN3146
Beilstein 4126243
Formula Weight 248.92
Physical Form Powder
Chemical Name or Material Di-n-butyltin oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.