missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-butyltin dilaurate, 95%, Thermo Scientific Chemicals
12.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Di-n-butyltin dilaurate is used as a catalyst for the production of polyurethanes as well as for the transesterification reactions. It is involved in the vulcanization of silicones and a stabilizer in polyvinyl chloride (PVC). It acts as a rust inhibitor for polyurethanes, polyols, silicones and as a fuel additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | <10°C |
Color | Yellow |
Density | 1.066 g/mL |
Boiling Point | >204°C (12 mmHg) |
Flash Point | 235°C (455°F) |
Linear Formula | [CH3(CH2)3]2Sn[O2C(CH2)10CH3]2 |
Refractive Index | 1.473 |
Quantity | 25 g |
UN Number | UN2788 |
Merck Index | 14,3038 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'