missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin dilaurate, 95%

Product Code. 11364819
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364819 500 g 500g
11386319 25 g 25g
11354819 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364819 Supplier Thermo Scientific Alfa Aesar Supplier No. 071130.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-58-7 | C32H64O4Sn

Di-n-butyltin dilaurate is used as a catalyst for the production of polyurethanes as well as for the transesterification reactions. It is involved in the vulcanization of silicones and a stabilizer in polyvinyl chloride (PVC). It acts as a rust inhibitor for polyurethanes, polyols, silicones and as a fuel additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-58-7
Molecular Formula C32H64O4Sn
MDL Number MFCD00008963
Synonym Di-n-butyltin didodecanoate

Specifications

Melting Point <10°C
Color Yellow
Density 1.066 g/mL
Boiling Point >204°C (12 mmHg)
Flash Point 235°C (455°F)
Linear Formula [CH3(CH2)3]2Sn[O2C(CH2)10CH3]2
Refractive Index 1.473
Quantity 500 g
UN Number UN2788
Merck Index 14,3038
Solubility Information Miscible with benzene and ether; Imiscible with water and methanol.
Formula Weight 631.55
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Di-n-butyltin dilaurate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Di-n-butyltin dilaurate, 95%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H360-May damage fertility or the unborn child.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Di-n-butyltin dilaurate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.