missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin diacetate, 95%, Thermo Scientific Chemicals

Catalog Number 11373447
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1067-33-0
C12H26O4Sn
353.05
MFCD00008697
LDZDQDDROJXUBO-UHFFFAOYSA-N
Diacetoxydi-n-butyltin
CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1067-33-0
C12H26O4Sn
353.05
MFCD00008697
LDZDQDDROJXUBO-UHFFFAOYSA-N
Diacetoxydi-n-butyltin
CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC

As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions

As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions.Di-n-butyltin diacetate is used as a stabilizer for polyvinyl chloride. It acts as a catalyst for the preparation of silicone and urethane foams. Further, it is used as a curing agent for silicone elastomers. In addition to this, it is used as a precursor for thin-film tin(IV) oxide gas sensing materials by laser-assisted chemical vapor deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 12°C
Color Colorless to Yellow
Density 1.32 g/mL
Boiling Point 142°C to 145°C (10 mmHg)
Flash Point 143°C (290°F)
Odor Acidic
Linear Formula [OOCCH3]2Sn[(CH2)3CH3]2
Refractive Index 1.47
Quantity 1 kg
UN Number UN2788
Solubility Information Soluble in water and most organic solvents; Miscible with most organic solvents.
Formula Weight 351.01
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Di-n-butyltin diacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'