missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin diacetate, 95%

Product Code. 11353447
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353447 250 g 250g
11363447 50 g 50g
11373447 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353447 Supplier Thermo Scientific Alfa Aesar Supplier No. 018595.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions | CAS: 1067-33-0 | C12H26O4Sn | 353.05 g/mol

As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions.Di-n-butyltin diacetate is used as a stabilizer for polyvinyl chloride. It acts as a catalyst for the preparation of silicone and urethane foams. Further, it is used as a curing agent for silicone elastomers. In addition to this, it is used as a precursor for thin-film tin(IV) oxide gas sensing materials by laser-assisted chemical vapor deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-33-0
Molecular Formula C12H26O4Sn
Molecular Weight (g/mol) 353.05
MDL Number MFCD00008697
InChI Key LDZDQDDROJXUBO-UHFFFAOYSA-N
Synonym Diacetoxydi-n-butyltin
SMILES CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC

Specifications

Melting Point 12°C
Color Colorless to Yellow
Density 1.32 g/mL
Boiling Point 142°C to 145°C (10 mmHg)
Flash Point 143°C (290°F)
Odor Acidic
Linear Formula [OOCCH3]2Sn[(CH2)3CH3]2
Refractive Index 1.47
Quantity 250 g
UN Number UN2788
Solubility Information Soluble in water and most organic solvents; Miscible with most organic solvents.
Formula Weight 351.01
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Di-n-butyltin diacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.