missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin bis(2-ethylhexanoate), Thermo Scientific Chemicals

Catalog Number 11344819
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2781-10-4
MFCD00015261
76044
This item is not returnable. View return policy
2781-10-4
MFCD00015261
76044

Di-n-butyltin bis(2-ethylhexanoate) is used in silanol condensation reactions for caulk and sealant applications and in the production of silicones. It is also used in one or two component systems which are compatible with aromatic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 57°C to 59°C
Density 1.070 g/mL
Boiling Point 220°C (2 mmHg)
Odor Odorless
Molecular Formula C22H21NO2S
Quantity 5 g
UN Number UN3146
Synonym s-trityl-l-cysteine, +-s-trityl-l-cysteine, h-cys trt-oh, s-tritylcysteine, tritylcysteine, 3-tritylthio-l-alanine, 2r-2-amino-3-triphenylmethyl sulfanyl propanoic acid, l-alanine, 3-tritylthio, stlc, s-triphenylmethyl-l-cysteine
Solubility Information Soluble in mineral oil, acetone, xylene
InChI Key DLMYFMLKORXJPO-UHFFFAOYNA-N
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)O)N
IUPAC Name (2R)-2-amino-3-tritylsulfanylpropanoic acid
Molecular Weight (g/mol) 363.48
Formula Weight 519.34
Physical Form Lumps
Chemical Name or Material Di-n-butyltin bis (2-ethylhexanoate)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'