missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin bis(2-ethylhexanoate)

Product Code. 11334819
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334819 100 g 100g
11344819 5 g 5g
11324819 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334819 Supplier Thermo Scientific Alfa Aesar Supplier No. 071126.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2781-10-4 | C24H48O4Sn | 519.35 g/mol

Di-n-butyltin bis(2-ethylhexanoate) is used in silanol condensation reactions for caulk and sealant applications and in the production of silicones. It is also used in one or two component systems which are compatible with aromatic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2781-10-4
MDL Number MFCD00015261

Specifications

Melting Point 57°C to 59°C
Density 1.070 g/mL
Boiling Point 220°C (2 mmHg)
Odor Odorless
Molecular Formula C24H48O4Sn
Quantity 100 g
UN Number UN3146
Synonym dibutyltin bis 2-ethylhexanoate, dibutyltin di 2-ethylhexoate, stannane, dibutylbis 2-ethyl-1-oxohexyl oxy, di-n-butyltin di-2-ethylhexanoate, di-n-butyltin bis 2-ethylhexanoate, dibutyltin di-2-ethylhexanoate, dibutyltin di 2-ethylhexanoate, dibutylbis 2-ethyl-1-oxohexyl oxy stannane, tin, dibutyl-, di 2-ethylhexoate, dibutyltin di-2-ethylhexoate
Solubility Information Soluble in mineral oil, acetone, xylene
InChI Key GPDWNEFHGANACG-UHFFFAOYNA-L
SMILES CCCCC(CC)C(=O)O[Sn](CCCC)(CCCC)OC(=O)C(CC)CCCC
IUPAC Name dibutyl[(2-ethylhexanoyl)oxy]stannyl 2-ethylhexanoate
Molecular Weight (g/mol) 519.35
Formula Weight 519.34
Physical Form Lumps
Chemical Name or Material Di-n-butyltin bis (2-ethylhexanoate)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.