missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin bis(2,4-pentanedionate), 95%

Product Code. 11436786
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436786 250 g 250g
11426786 50 g 50g
11416786 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436786 Supplier Thermo Scientific Alfa Aesar Supplier No. B23921.30
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22673-19-4 | C18H32O4Sn

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22673-19-4
Molecular Formula C18H32O4Sn
MDL Number MFCD00077994
Synonym dibutyltin bis acetylacetonate, di-n-butylbis 2,4-pentanedionate tin, dibutyltin bis 2,4-pentanedionate, 3-penten-2-one, 4,4'-dibutylstannylene bis oxy bis-, 3z,3'z, 3z-4-dibutyl 2z-4-oxopent-2-en-2-yl oxy stannyl oxy pent-3-en-2-one, z-4-dibutyl-z-1-methyl-3-oxo-but-1-enoxy stannyl oxypent-3-en-2-one

Specifications

Melting Point 26°C
Density 1.22
Boiling Point 150°C
Flash Point 40°C (104°F)
Odor Weak
Refractive Index 1.515
Quantity 250 g
UN Number UN1992
Formula Weight 431.16
Percent Purity 95%
Chemical Name or Material Di-n-butyltin bis(2,4-pentanedionate)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.