missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl methylphosphonate, Thermo Scientific™

Catalog Number 11327278
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
2404-73-1
C9H21O3P
208.238
MFCD00015242
NPNBLTJGRBYCJB-UHFFFAOYSA-N
dibutyl methylphosphonate, dibutyl methanephosphonate, di-n-butyl methylphosphonate, dibutylmethylphosphonate, di-n-butyl methanephosphonate, methylphosphonic acid dibutyl ester, methylphosphonic acid, dibutyl ester, 1-butoxy methyl phosphoryl oxybutane, phosphonic acid, methyl-, dibutyl ester, phosphonic acid, methyl-, o,o-dibutyl ester
16994
1-[butoxy(methyl)phosphoryl]oxybutane
CCCCOP(=O)(C)OCCCC
This item is not returnable. View return policy
2404-73-1
C9H21O3P
208.238
MFCD00015242
NPNBLTJGRBYCJB-UHFFFAOYSA-N
dibutyl methylphosphonate, dibutyl methanephosphonate, di-n-butyl methylphosphonate, dibutylmethylphosphonate, di-n-butyl methanephosphonate, methylphosphonic acid dibutyl ester, methylphosphonic acid, dibutyl ester, 1-butoxy methyl phosphoryl oxybutane, phosphonic acid, methyl-, dibutyl ester, phosphonic acid, methyl-, o,o-dibutyl ester
16994
1-[butoxy(methyl)phosphoryl]oxybutane
CCCCOP(=O)(C)OCCCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 2 g
UN Number UN3272
Formula Weight 208.24
Physical Form Liquid
Chemical Name or Material Di-n-butyl methylphosphonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'