missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl carbonate

Product Code. 16564254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16564254 25 g 25g
16554254 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16564254 Supplier Thermo Scientific Acros Supplier No. 463880250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 542-52-9 | C9H18O3 | 174.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-52-9
Molecular Formula C9H18O3
Molecular Weight (g/mol) 174.24
InChI Key QLVWOKQMDLQXNN-UHFFFAOYSA-N
IUPAC Name dibutyl carbonate
SMILES CCCCOC(=O)OCCCC

Specifications

Density 0.9200 g/mL
Quantity 25 g
Specific Gravity 0.92
Formula Weight 174.24
Chemical Name or Material Di-n-butyl carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.