missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl carbonate

Product Code. 16554254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16554254 5 g 5g
16564254 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16554254 Supplier Thermo Scientific Acros Supplier No. 463880050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 542-52-9 | C9H18O3 | 174.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-52-9
Molecular Formula C9H18O3
Molecular Weight (g/mol) 174.24
InChI Key QLVWOKQMDLQXNN-UHFFFAOYSA-N
IUPAC Name dibutyl carbonate
SMILES CCCCOC(=O)OCCCC

Specifications

Density 0.9200 g/mL
Quantity 5 g
Specific Gravity 0.92
Formula Weight 174.24
Chemical Name or Material Di-n-butyl carbonate
compliance-icons compliance-icons
Product Identifier
  • Di-n-butyl carbonate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Carbonic acid, dibutyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.