missing translation for 'onlineSavingsMsg'
Learn More

Di-N-butyl Amidosulfenyl Chloride, TRC

Product Code. 30384988
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384988 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384988 Supplier Toronto Research Chemicals Supplier No. TRCD42944050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-N-butyl Amidosulfenyl Chloride
CAS 6541-82-8
Molecular Formula C8 H18 Cl N S
Quantity 50 mg
Synonym N,N-Dibutyl-amidosulfenyl Chloride, Dibutylaminosulfur Chloride, Dibutylamidosulfenyl chloride, Chloro Dibutylamino Sulfide
InChI Formula InChI=1S/C8H18ClNS/c1-3-5-7-10(11-9)8-6-4-2/h3-8H2,1-2H3
SMILES CCCCN(CCCC)SCl
IUPAC Name (dibutylamino) thiohypochlorite
Molecular Weight (g/mol) 195.753
Formula Weight 195.085
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.