missing translation for 'onlineSavingsMsg'
Learn More

Di-Mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(Ii)] Chloride, TRC

Código de producto. 30377786
Cambiar vista
Click to view available options
Quantity:
100 mg
Tamaño de la unidad:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
30377786 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30377786 Proveedor Toronto Research Chemicals N.º de proveedor TRCD460385100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-Mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(Ii)] Chloride
CAS 30698-64-7
Molecular Formula C12 H34 Cu2 N4 O2
Quantity 100 mg
InChI Formula InChI=1S/2C6H16N2.2Cu.2H2O/c2*1-7(2)5-6-8(3)4;;;;/h2*5-6H2,1-4H3;;;2*1H2/q;;2*+2;;/p-2
SMILES C[N]1(C)CC[N](C)(C)[Cu+2]12[OH-][Cu+2]3([OH-]2)[N](C)(C)CC[N]3(C)C
Molecular Weight (g/mol) 393.516
Formula Weight 392.126
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.