missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Methyl-Histone H3 (Lys36) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29202-1-AP-20UL
This item is not returnable.
View return policy
Description
The Di-Methyl-Histone H3 (Lys36) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Di-Methyl-Histone H3 (Lys36). This antibody recognizes human, mouse, rat antigen. The Di-Methyl-Histone H3 (Lys36) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| Di-Methyl-Histone H3 (Lys36) | |
| Polyclonal | |
| Unconjugated | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 15-18 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8350 | |
| Store at -20°C. | |
| Polyclonal Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction