missing translation for 'onlineSavingsMsg'
Learn More

Di-^m-chlorobis(norbornadiene)dirhodium(I), Rh 44% min

Product Code. 11395968
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
This item is not returnable. View return policy

Product Code. 11395968

Brand: Thermo Scientific Alfa Aesar 010467.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12257-42-0 | C14H16Cl2Rh2 | 460.99 g/mol

Di-μ-chlorobis(norbornadiene)dirhodium(I) is used as catalysts in olefin hydroformylation, olefin hydrogenation and olefin isomerization

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12257-42-0
Molecular Formula C14H16Cl2Rh2
Molecular Weight (g/mol) 460.99
MDL Number MFCD00198060
InChI Key RXDWVIOULPVOEO-UHFFFAOYSA-L
Synonym bicyclo 2.2.1 hepta-2,5-diene-rhodium chloride dimer, bicyclo 2.2.1 hepta-2,5-diene; rhodium 2+ ; dichloride, bis 2,3,5,6-eta-bicyclo 2.2.1 hepta-2,5-diene di-mu-chlorodirhodium
PubChem CID 114600
SMILES [Cl-].[Cl-].[Rh+].[Rh+].C1C2C=CC1C=C2.C1C2C=CC1C=C2

Specifications

Odor Odorless
Quantity 1 g
Solubility Information Slightly soluble in acetone,chloroform,methanol and insoluble in water.
IUPAC Name λ²-rhodium(2+) bis(bicyclo[2.2.1]hepta-2,5-diene) dichloride
Formula Weight 461.01
Concentration or Composition (by Analyte or Components) Rh 44% min.
Physical Form Powder
Chemical Name or Material Di-mu-chlorobis(norbornadiene)dirhodium(I)

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato