missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ DI Liners for Shimadzu GCs
Description
- Trace, dirty, high MW, active samples, high recovery and linearity
- Geometry: Uniliner
- Material: Borosilicate Glass
- Deactivation: Intermediate Polarity (IP)
- Packing: Deactivated Wool
- I.D.: 3.5mm
- OD: 5mm
- Length: 95mm
Specifications
Specifications
| Quantity | 5/Pk. |
| Length (Metric) | 9.5 cm |
| Packing Material | Deactivated Wool |
| Product Type | IP Uniliner Inlet Liner |
| Diameter (Metric) Inner | 0.35 cm |
| Diameter (Metric) Outer | 0.5 cm |
| For Use With (Equipment) | Shimadzu 17 A/2010/2014 GCs w/ Split/Splitless Inlets |
| Liner Material | Borosilicate Glass |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?