missing translation for 'onlineSavingsMsg'
Learn More

Di-Iso-Propyl Ether, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical™

Click to view available options
Quantity:
1L
2.5L
5L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Plastic container
This item is not returnable. View return policy
108-20-3
C6H14O
102.177
8880
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
7914
2-propan-2-yloxypropane
CC(C)OC(C)C
This item is not returnable. View return policy
108-20-3
C6H14O
102.177
8880
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
7914
2-propan-2-yloxypropane
CC(C)OC(C)C
Color Colorless
Melting Point -85.5°C
Boiling Point 68°C
Packaging Amber glass bottle
Quantity 1L
Formula Weight 102.18g/mol
Physical Form Liquid
Vapor Density 1.42
compliance-icons compliance-icons
Product Identifier
  • Isopropyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Supplemental information
  • MIXTURE LIST-Contains : Isopropyl ether, 2,6-Di-tert-butyl-p-cresol