missing translation for 'onlineSavingsMsg'
Learn More

Di-Iodomethane, Technical, SLR, Fisher Chemical

Product Code. 10348090 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Amber glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10348090 25 mL Amber glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10348090 Supplier Fisher Chemical Supplier No. D/3200/04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75-11-6
Melting Point 6°C
Color Orange to Yellow
Boiling Point 181°C
Packaging Amber glass bottle
Molecular Formula CH2I2
MDL Number MFCD00001079
Quantity 25 mL
Synonym methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
InChI Key NZZFYRREKKOMAT-UHFFFAOYSA-N
SMILES ICI
IUPAC Name diiodomethane
Molecular Weight (g/mol) 267.84
PubChem CID 6346
Vapor Density 9.25
Formula Weight 267.83g/mol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diiodomethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.