missing translation for 'onlineSavingsMsg'
Learn More

Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride, 98%

Product Code. 10081701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081701 5 g Glass bottle 5g
10470242 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081701 Supplier Thermo Scientific Acros Supplier No. 348980050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30698-64-7 | C12H36Cl2Cu2N4O2 | 466.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30698-64-7
Molecular Formula C12H36Cl2Cu2N4O2
Molecular Weight (g/mol) 466.44
MDL Number MFCD01321163
InChI Key VVXKYYDFGPZSOZ-UHFFFAOYSA-L
Synonym cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
PubChem CID 11190467
IUPAC Name chlorocopper;N,N,N',N'-tetramethylethane-1,2-diamine;dihydrate
SMILES O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C

Specifications

Melting Point 136.0°C to 138.0°C
Color Purple
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 464.42
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Di-μ-hydroxo-bis-[(N,N,N′,N′-tetramethylethylenediamine)copper(II)]chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.