missing translation for 'onlineSavingsMsg'
Learn More

Di-Beta,Beta'-Chloroethylphosphoric Acid, TRC

Product Code. 17994035
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17994035 250 mg 250mg
17924115 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17994035 Supplier Toronto Research Chemicals Supplier No. TRCC366790250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3040-56-0
Molecular Formula C4H9Cl2O4P
Molecular Weight (g/mol) 222.99
Synonym Ethanol, 2-chloro-, hydrogen phosphate (7CI, 8CI, 9CI), Bis(2-chloroethyl) hydrogen phosphate, Bis(2-chloroethyl) phosphate, Bis(chloroethyl) phosphate, Bis(β-chloroethyl) phosphate, Bis(β-chloroethyl) phosphoric acid, Di-2-Chloroethyl phosphate, Di-β,β'-Chloroethylphosphoric acid
IUPAC Name bis(2-chloroethyl) hydrogen phosphate
SMILES OP(=O)(OCCCl)OCCCl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C4H9Cl2O4P/c5-1-3-9-11(7,8)10-4-2-6/h1-4H2,(H,7,8)
Formula Weight 221.9616
Percent Purity >95%
Chemical Name or Material Di-beta,beta-Chloroethylphosphoric Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.