missing translation for 'onlineSavingsMsg'
Learn More

Di-2-thienyl sulfide, 97%

Product Code. 15446097
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446097 1 g 1g
15456097 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446097 Supplier Thermo Scientific Alfa Aesar Supplier No. H33284.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3988-99-6 | C8H6S3 | 198.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3988-99-6
Molecular Formula C8H6S3
Molecular Weight (g/mol) 198.32
MDL Number MFCD00052047
InChI Key AKYIWBKPINOXJY-UHFFFAOYSA-N
Synonym 2-2-thienylthio thiophene, thiophene, 2,2'-thiobis, thiophene,2,2'-thiobis, 2-2-thienylsulfanyl thiophene, 2-thiophen-2-ylsulfanyl thiophene, 2-thienyl sulfide, di-2-thienyl sulfide, 2,2'-thiobisthiophene, di 2-thienyl sulphide
PubChem CID 138094
IUPAC Name 2-thiophen-2-ylsulfanylthiophene
SMILES S(C1=CC=CS1)C1=CC=CS1

Tekniske data

Density 1.311
Boiling Point 102°C to 104°C (0.9 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.649
Quantity 1 g
Formula Weight 198.32
Percent Purity 97%
Chemical Name or Material Di-2-thienyl sulfide

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.