missing translation for 'onlineSavingsMsg'
Learn More

Di-2-pyridyl thionocarbonate, 98%

Product Code. 11440617
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440617 25 g 25g
11420617 1 g 1g
11430617 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440617 Supplier Thermo Scientific Alfa Aesar Supplier No. B22451.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96989-50-3 | C11H8N2O2S | 232.257 g/mol

Di-2-pyridyl thionocarbonate is used in the synthesis of 10-des(carbamoyloxy)-10-isothiocyanatoporfiromycin. It is also used as a substitute of thiophosgene, in the preparation of isothiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-2-pyridyl thionocarbonate is used in the synthesis of 10-des(carbamoyloxy)-10-isothiocyanatoporfiromycin. It is also used as a substitute of thiophosgene, in the preparation of isothiocyanate.

Solubility
Soluble in water 9280 g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 96989-50-3
Molecular Formula C11H8N2O2S
Molecular Weight (g/mol) 232.257
MDL Number MFCD00074870
InChI Key IKYOVSVBLHGFMA-UHFFFAOYSA-N
Synonym di-2-pyridyl thionocarbonate, o,o-di pyridin-2-yl carbonothioate, o,o'-di-2-pyridyl thiocarbonate, o,o-dipyridin-2-yl carbonothioate, di 2-pyridyl thionocarbonate, carbonothioic acid, o,o-di-2-pyridinyl ester, di 2-pyridyloxy methane-1-thione, o,o-dipyridin-2-yl thiocarbonate, bis pyridin-2-yloxy methanethione, di-2-pyridyl thiocarbomate
PubChem CID 719784
IUPAC Name dipyridin-2-yloxymethanethione
SMILES C1=CC=NC(=C1)OC(=S)OC2=CC=CC=N2

Spezifikation

Melting Point 96°C to 99°C
Quantity 25 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water 9280g/L).
Formula Weight 232.26
Percent Purity 98%
Chemical Name or Material Di-2-pyridyl thionocarbonate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.