missing translation for 'onlineSavingsMsg'
Learn More

Di(2-pyridyl) ketone, 98%

Product Code. 15421548
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421548 25 g 25g
15401548 1 g 1g
15411548 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421548 Supplier Thermo Scientific Alfa Aesar Supplier No. H55322.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19437-26-4 | C11H8N2O | 184.198 g/mol

di-2-Pyridyl ketone has been used in the preparation of unusual {Ni(II)(3)Ln(III)(?-OR)(6)}(3+) complexes with a star topology. Its monoanionic form has been used in the synthesis of triangular Ni(2)M (M = lanthanide, Y) complexes. The Schiff base ligands derived from di-2-pyridyl ketone exhibit ant proliferative activity in SK-N-MC neuroepithelioma (cancer) cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
di-2-Pyridyl ketone has been used in the preparation of unusual {Ni(II)(3)Ln(III)(μ-OR)(6)}(3+) complexes with a ″star″ topology. Its monoanionic form has been used in the synthesis of triangular Ni(2)M (M = lanthanide, Y) complexes. The Schiff base ligands derived from di-2-pyridyl ketone exhibit ant proliferative activity in SK-N-MC neuroepithelioma (cancer) cells.

Solubility
Low solubility in water.

Notes
Desiccate at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19437-26-4
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.198
MDL Number MFCD00006288
InChI Key QPOWUYJWCJRLEE-UHFFFAOYSA-N
Synonym di-2-pyridyl ketone, di pyridin-2-yl methanone, bis 2-pyridyl ketone, 2,2'-carbonyldipyridine, di 2-pyridyl ketone, methanone, di-2-pyridinyl, 2,2'-dipyridyl ketone, 2-pyridine-2-carbonyl pyridine, pyridyl ketone
PubChem CID 88065
IUPAC Name dipyridin-2-ylmethanone
SMILES C1=CC=NC(=C1)C(=O)C2=CC=CC=N2

Specifications

Melting Point 52°C to 55°C
Flash Point 110°C (230°F)
Quantity 25 g
Beilstein 126598
Solubility Information Low solubility in water.
Formula Weight 184.2
Percent Purity 99%
Chemical Name or Material Di(2-pyridyl) ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.