missing translation for 'onlineSavingsMsg'
Learn More

Di-(2-HydroxypHenylimino)Ethane, Pure, for Calcium Determination, Fisher Chemical

Product Code. 10338090 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Plastic powder jar
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10338090 10 g Plastic powder jar 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10338090 Supplier Fisher Chemical Supplier No. D/3100/45

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1149-16-2
Melting Point 201°C
Color Beige
Packaging Plastic powder jar
Molecular Formula C14H12N2O2
MDL Number MFCD00002183
Quantity 10 g
Synonym glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
InChI Key OMJNMRJMNUREOJ-KOBPNRPCSA-N
SMILES OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O
IUPAC Name 6-[(E)-2-(2-hydroxyanilino)ethenyl]iminocyclohexa-2,4-dien-1-one
Molecular Weight (g/mol) 240.26
PubChem CID 5356911
Formula Weight 240.26g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • Di-(2-hydroxyphenylimino)ethane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.